Amphotericin B solubilized
Synonym(s):
Amphotericin B from Streptomyces sp., Fungizone, Amphotericin B – Low Endotoxin
Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Quality Segment
Mode of Action
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing