Skip to Content
Merck
Rapamycin from Streptomyces hygroscopicus ≥95% (HPLC), powder

Rapamycin

Rapamycin from Streptomyces hygroscopicus ≥95% (HPLC), powder
Synonym(s):
Rapamycin, 23,27-Epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine, AY 22989, Sirolimus
Empirical Formula (Hill Notation):
C51H79NO13
CAS Number:
Molecular Weight:
914.17

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing