Skip to Content
CH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Folic acid
Synonym(s):
PteGlu, Pteroyl-L-glutamic acid, Vitamin M
Empirical Formula (Hill Notation):
C
19
H
19
N
7
O
6
CAS Number:
59-30-3
Molecular Weight:
441.40
EC Number:
200-419-0
Products
Product Category
vitamins (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
F7876
Folic acid,
≥97%
Expand
View Pricing
F8758
Folic acid,
BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Expand
View Pricing
F8798
Folic acid,
meets USP testing specifications
Expand
View Pricing