Skip to Content
Merck
Sodium Phosphate OmniPur® Grade, >= 98.0%

Sodium phosphate dibasic heptahydrate

Sodium Phosphate OmniPur® Grade, >= 98.0%
Synonym(s):
Disodium hydrogen phosphate heptahydrate, Disodium phosphate, Sodium phosphate dibasic heptahydrate
Linear Formula:
Na2HPO4 · 7H2O
CAS Number:
Molecular Weight:
268.07

Products

Product Category
Brand
Color
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade