Skip to Content
Merck
Aluminum chloride anhydrous, sublimed, ≥98%

Aluminum chloride

Aluminum chloride anhydrous, sublimed, ≥98%
Synonym(s):
Aluminium chloride
Linear Formula:
AlCl3
CAS Number:
Molecular Weight:
133.34
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Core
Special Grade