Skip to Content
CH
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Agarose
Synonym(s):
OmniPur
®
Agarose, Agarose, OmniPur
®
Agarose, Low Melting, Agarose, Type C
CAS Number:
9012-36-6
EC Number:
232-731-8
Products
Product Category
carbohydrates (17)
Brand
Millipore (6)
Sigma-Aldrich (16)
Biological Source
Color
Feature
Melting Point (°C)
Physical Form
Research Area
Quality Segment
Technique
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
A9539
Agarose,
BioReagent, for molecular biology, low EEO
Expand
View Pricing
A6013
Agarose,
Type I, low EEO
Expand
View Pricing
A0576
Agarose,
Low EEO
Expand
View Pricing
05066
Agarose,
High EEO, for molecular biology
Expand
View Pricing
A4718
Agarose,
for molecular biology
Expand
View Pricing
2120-OP
Agarose,
OmniPur
®
Grade
Expand
View Pricing
A4679
Agarose,
Low EEO, for Immunoelectrophoresis
Expand
View Pricing
A2576
Agarose,
Ultra-low Gelling Temperature, molecular biology grade
Expand
View Pricing
A5030
Agarose,
Ultra-low Gelling Temperature
Expand
View Pricing
1.16802
Agarose,
(low electroendoosmosis) for electrophoresis
Expand
View Pricing
A6877
Agarose,
Medium EEO, for molecular biology
Expand
View Pricing
A0169
Agarose,
Type I-A, low EEO
Expand
View Pricing
1.01236
Agarose,
for analytical nucleic acid electrophoresis
Expand
View Pricing
A5304
Agarose,
Special High EEO
Expand
View Pricing
A2929
Agarose,
For pulsed field electrophoresis running gel
Expand
View Pricing
2070-OP
Agarose,
OmniPur
®
Grade, Low Melting
Expand
View Pricing
1.16801
Agarose,
(medium electroendoosmosis) for electrophoresis
Expand
View Pricing
A9793
Agarose,
High EEO
Expand
View Pricing
A9918
Agarose,
Type II-A, Medium EEO
Expand
View Pricing
A2790
Agarose,
Wide range, for molecular biology
Expand
View Pricing
Page 1 of 2
Page 1 of 2