Skip to Content
Merck
ECO TWEEN® 80 viscous liquid

TWEEN® 80

ECO TWEEN® 80 viscous liquid
Synonym(s):
TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80
CAS Number:

Products

Product Category
Brand
Sustainability
Biological Source
Feature
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Application
Special Grade
Technique