Skip to Content
Merck
Methyl cellulose 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP

Methyl cellulose

Methyl cellulose 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Physical Form
Product Line
Agency
Tissue
Quality Segment
Application
Technique