Skip to Content
Merck
Cyclosporin A from Tolypocladium inflatum, BioReagent, for molecular biology, ≥95%

Cyclosporin A

Cyclosporin A from Tolypocladium inflatum, BioReagent, for molecular biology, ≥95%
Synonym(s):
Antibiotic S 7481F1, Cyclosporine
Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Technique