Skip to Content
Merck
All Photos(1)

Documents

305839

Sigma-Aldrich

3-Dimethylaminoacrolein

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH=CHCHO
CAS Number:
Molecular Weight:
99.13
Beilstein:
969389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

liquid

refractive index

n20/D 1.584 (lit.)

bp

270-273 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

CN(C)\C=C\C=O

InChI

1S/C5H9NO/c1-6(2)4-3-5-7/h3-5H,1-2H3/b4-3+

InChI key

RRLMPLDPCKRASL-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Dimethylaminoacrolein was used in synthesis of:
  • benzochlorins, benzoisobacteriochlorins and benzobacteriochlorins
  • styryl-substituted Z/E-chlorin derivatives with chlorophyll-a skeleton
  • Y shaped cyanines having dimethylamino end groups

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient new syntheses of benzochlorins, benzoisobacteriochlorins, and benzobacteriochlorins.
Graca M, et al.
Tetrahedron Letters, 31(10), 1365-1368 (1990)
Triple-branched cyanine dyes.
Dale J and Eriksen OI.
Acta Chemica Scandinavica. Series B, 42(4), 242-246 (1988)
Synthesis of Styryl-substituted Z/E-Chlorin Derivatives with Chlorophyll-a Skeleton
Wang J-J, et al.
Chin. J. Chem., 24(7), 933-938 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service