MilliporeSigma
Sexually Dimorphic Expression of Reelin in the Brain of a Mouse Model of Alzheimer Disease.
Journal of molecular neuroscience : MN (2016-11-21)
Giampiero Palladino, Vincenzina Nicolia, Gabor G Kovacs, Sonia Canterini, Viviana Ciraci, Andrea Fuso, Franco Mangia, Sigfrido Scarpa, Maria Teresa Fiorenza
RÉSUMÉ
Recent evidence highlights the protective role of reelin against amyloid β (Aβ)-induced synaptic dysfunction and cognitive impairment in Alzheimer disease (AD). In this study, exploiting TgCRND8 mice that overexpress a mutant form of amyloid β precursor protein (AβPP) and display an early onset of AD neuropathological signs, we addressed the question whether changes of reelin expression eventually precede the appearance of Aβ-plaques in a sex-dependent manner. We show that sex-associated and brain region-specific differences in reelin expression appear long before Aβ-plaque formation. However, in spite of a downregulation of reelin expression compared to males, TgCRND8 females display fewer Aβ-plaques, suggesting that additional factors, other than sex and reelin level, influence amyloidosis in this mouse model.