MilliporeSigma
  • Sevelamer does not decrease lipopolysaccharide or soluble CD14 levels but decreases soluble tissue factor, low-density lipoprotein (LDL) cholesterol, and oxidized LDL cholesterol levels in individuals with untreated HIV infection.
Sevelamer does not decrease lipopolysaccharide or soluble CD14 levels but decreases soluble tissue factor, low-density lipoprotein (LDL) cholesterol, and oxidized LDL cholesterol levels in individuals with untreated HIV infection.
The Journal of infectious diseases (2014-05-28)
Netanya G Sandler, Xinyan Zhang, Ronald J Bosch, Nicholas T Funderburg, Andrew I Choi, Janet K Robinson, Derek M Fine, Robert W Coombs, Jeffrey M Jacobson, Alan L Landay, Daniel C Douek, Randall Tressler, Sarah W Read, Cara C Wilson, Steven G Deeks, Michael M Lederman, Rajesh T Gandhi
RÉSUMÉ
Abnormal levels of inflammation are associated with cardiovascular disease and mortality in human immunodeficiency virus (HIV)-infected patients. Microbial translocation, which may cause inflammation, is decreased by sevelamer in patients undergoing hemodialysis. In this single-arm study, we evaluated the effects of 8 weeks of sevelamer therapy on 36 HIV-infected subjects who were not receiving antiretroviral therapy. Sevelamer did not significantly change markers of microbial translocation, inflammation, or T-cell activation. During sevelamer treatment, however, levels of soluble tissue factor, low-density lipoprotein (LDL) cholesterol, and oxidized LDL cholesterol decreased significantly, whereas D-dimer levels increased. Thus, in this study population, sevelamer did not reduce microbial translocation but may have yielded cardiovascular benefits. NCT 01543958.