MilliporeSigma
Mutations in GPC3, a glypican gene, cause the Simpson-Golabi-Behmel overgrowth syndrome.
Nature genetics (1996-03-01)
G Pilia, R M Hughes-Benzie, A MacKenzie, P Baybayan, E Y Chen, R Huber, G Neri, A Cao, A Forabosco, D Schlessinger
RÉSUMÉ
Simpson-Golabi-Behmel syndrome (SGBS) is an X-linked condition characterized by pre- and postnatal overgrowth with visceral and skeletal anomalies. To identify the causative gene, breakpoints in two female patients with X;autosome translocations were identified. The breakpoints occur near the 5' and 3' ends of a gene, GPC3, that spans more than 500 kilobases in Xq26; in three families, different microdeletions encompassing exons cosegregate with SGBS. GPC3 encodes a putative extracellular proteoglycan, glypican 3, that is inferred to play an important role in growth control in embryonic mesodermal tissues in which it is selectively expressed. Initial western- and ligand-blotting experiments suggest that glypican 3 forms a complex with insulin-like growth factor 2 (IGF2), and might thereby modulate IGF2 action.