MilliporeSigma
Attenuation of acute inflammatory response by atorvastatin after spinal cord injury in rats.
Journal of neuroscience research (2004-12-18)
Ravinder Pannu, Ernest Barbosa, Avtar K Singh, Inderjit Singh
RÉSUMÉ
Spinal cord injury (SCI) is a devastating and complex clinical condition involving proinflammatory cytokines and nitric oxide toxicity that produces a predictable pattern of progressive injury entailing neuronal loss, axonal destruction, and demyelination at the site of impact. The involvement of proinflammatory cytokines and inducible nitric oxide synthase (iNOS) in exacerbation of SCI pathology is well documented. We have reported previously the antiinflammatory properties and immunomodulatory activities of statins (3-hydroxy-3-methylglutaryl [HMG]-CoA reductase inhibitors) in the animal model of multiple sclerosis, experimental allergic encephalitis (EAE). The present study was undertaken to investigate the efficacy of atorvastatin (Lipitor; LP) treatment in attenuating SCI-induced pathology. Immunohistochemical detection and real-time PCR analysis showed increased expression of iNOS, tumor necrosis factor alpha (TNFalpha) and interleukin 1beta (IL-1beta) after SCI. In addition, neuronal apoptosis was detected 24 hr after injury followed by a profound increase in ED1-positive inflammatory infiltrates, glial fibrillary acidic protein (GFAP)-positive reactive astrocytes, and oligodendrocyte apoptosis by 1 week after SCI relative to control. LP treatment attenuated the SCI-induced iNOS, TNFalpha, and IL-1beta expression. LP also provided protection against SCI-induced tissue necrosis, neuronal and oligodendrocyte apoptosis, demyelination, and reactive gliosis. Furthermore, rats treated with LP scored much higher on the locomotor rating scale after SCI (19.13 +/- 0.53) than did untreated rats (9.04 +/- 1.22). This study therefore reports the beneficial effect of atorvastatin for the treatment of SCI-related pathology and disability.