MilliporeSigma
Diverse roles of the nucleic acid-binding protein KHSRP in cell differentiation and disease.
Wiley interdisciplinary reviews. RNA (2015-12-29)
Paola Briata, Domenico Bordo, Margherita Puppo, Franco Gorlero, Martina Rossi, Nora Perrone-Bizzozero, Roberto Gherzi
RÉSUMÉ
The single-stranded nucleic acid-binding protein KHSRP (KH-type splicing regulatory protein) modulates RNA life and gene expression at various levels. KHSRP controls important cellular functions as different as proliferation, differentiation, metabolism, and response to infectious agents. We summarize and discuss experimental evidence providing a potential link between changes in KHSRP expression/function and human diseases including neuromuscular disorders, obesity, type II diabetes, and cancer.