Mixers for Bioprocessing
Mobius® single-use mixing solutions deliver efficient mixing of pharmaceutical ingredients from intermediate to final drug products, as well as preparation of process solutions such as buffers and media. Unlike traditional stainless steel mixers, single-use mixers reduce downtime spent on process steps such as cleaning in place (CIP), sterilization in place (SIP), cleaning validation, and process engineering. Mobius® single-use mixing solutions are capable of speeds up to 1000 RPM, meeting the broadest range of mixing production requirements. Mobius® flexibility offers you a choice of just the right filter, connector, and tubing options for your specific application and environment, including sterile interfaces, from fermentation to final fill.
Mobius® single-use mixing solutions include a complete range of mixers in sizes from 10 L to 3,000 L. Both Mobius® MIX and Mobius® Power MIX product lines offer proven operational efficiency, saving valuable processing and validation time. Our scalable, ready-to-use systems and processing technologies reduce the risk of contamination and deliver reproducible results.
- Easy setup and use
- Closed, zero dead-leg sterile sampling system enables representative samples
- Single-use technology for increased production capacity in multiproduct facilities
- Validation, quality, and regulatory compliance support
Mobius® MIX: the mixing solution of choice for gentle mixing applications such as protein and shear-sensitive product homogenization
The Mobius® MIX family of single-use mixers is offered in sizes from 10 L to 1,000 L. Each single-use mixing container includes our proprietary, magnetically driven levitating impeller for improved mixing consistency and efficiency.
- Available in 10 L, 50 L, 100 L, 200 L, 500 L, and 1,000 L sizes
- Fast, easy setup and use
- Compact footprint and mobile carrier construction for enhanced flexibility
- Instant-on electronics with no warmup needed
Mobius® Power MIX: the mixing solution of choice for hard-to-dissolve materials and buffer preparation
The Mobius® Power MIX family of single-use mixers combines high-performance mixing technology with design features that make it easy to use. The impeller design and motor are based on our magnetically coupled NovAseptic® mixing technology, a proven mixing technology used with stainless steel tanks for efficient mixing of the most challenging buffer, media, and biopharmaceutical ingredients.
- Available in a wide range of sizes and materials
- 100 L, 200 L, 500 L (HDPE and jacketed stainless steel)
- 1,000 L, 2,000 L, 2,500 L, and 3,000 L (jacketed stainless steel only)
- Stainless steel versions have wide doors that open to one half the mixer's diameter for easy bag loading
- Options on the stainless steel versions include integrated load cells and temperature sensor
Related Product Resources
Application Note: Mobius® Power MIX: Scalability in Mixing
Application Note: Mobius® Power Mix 500
Spec Sheet: Mobius® Power MIX 100, 200 and 500 Single-Use Mixers
Spec Sheet: Mobius® Power MIX 2000 Single-Use Mixer
Specification Sheet: Mobius® Power MIX 1000, 2000, 2500 and 3000 Single-Use Mixers