Principaux documents
TT38440SG-EP
Eppendorf twin.tec® PCR Plates 384
skirted, PCR clean, volume 40 μL, colorless wells, green plate, pack of 25
About This Item
Code UNSPSC :
41106300
Matériaux:
colorless wells
green plate
polypropylene wells
Volume:
40 μL
Volume de travail:
40 μL
Stérilité:
non-sterile
Caractéristiques:
skirt
PCR clean
skirted
Matériaux
colorless wells
green plate
polypropylene wells
Taille des puits
384
Stérilité
non-sterile
Caractéristiques
skirt
PCR clean
skirted
Conditionnement
pack of 25
Fabricant/nom de marque
Eppendorf® 951020729 (NA)
Technique(s)
PCR: suitable
Volume
40 μL
Volume de travail
40 μL
Catégories apparentées
Description générale
The Eppendorf twin.tec PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
Caractéristiques et avantages
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler X50 and Mastercycler nexus
  • 40 µL max. well volume
  • Ideal for quantitative real-time PCR as well – OptiTrack matrix for faster sample identification and fewer pipetting errors
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG