Key Documents
SRP6359
Sigma-Aldrich
FGF14 human
recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Synonyme(s) :
FGF-14, FHF-4, FHF4, Fibroblast growth factor 14 isoform 1A, SCA27
About This Item
Code UNSPSC :
12352202
Source biologique
human
Produit recombinant
expressed in E. coli
Pureté
≥90% (SDS-PAGE)
Forme
liquid
Poids mol.
30 kDa (271 aa, 1-247 aa + His Tag)
Conditionnement
pkg of 50 μg
Concentration
0.25 mg/mL
Numéro d'accès UniProt
Conditions d'expédition
dry ice
Température de stockage
−70°C
Informations sur le gène
human ... FGF14(2259)
Description générale
FGF14 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. A mutation in this gene is associated with autosomal dominant cerebral ataxia. Alternatively spliced transcript variants have been found for this gene. Recombinant human FGF14 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.
Forme physique
0.25 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.2 M NaCl, 5 mM DTT and 50% glycerol.
Pictogrammes
Exclamation mark
Mention d'avertissement
Warning
Mentions de danger
Classification des risques
Eye Irrit. 2
Code de la classe de stockage
10 - Combustible liquids
Classe de danger pour l'eau (WGK)
WGK 3
Point d'éclair (°F)
Not applicable
Point d'éclair (°C)
Not applicable