Key Documents
Z648434
Quickfit® bends with vacuum connection
socket joint: ST/NS 19/26, cone joint: ST/NS 19/26
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
cone joint: ST/NS19/26
socket joint: ST/NS19/26
Fabricant/nom de marque
Quickfit RA 3/22/SC
Disponibilité
not available in USA and Canada
not available in USA and Canada
Description générale
Socket to cone. With screw-thread adapter for flexible tubing.
Informations légales
Quickfit is a registered trademark of SciLabware Limited