Key Documents
TEM-LC300NIUL
Sigma-Aldrich
Ultrathin Lacey Carbon Supported Nickel Grid
size 300 mesh, box of 25
Synonyme(s) :
LACEY CARBON FILM - NI 300 MESH ULTRA THIN 25/BX
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 25
Taille
300 mesh
Catégories apparentées
Description générale
Ultrathin Lacey Carbon (4-6 nm) Supported Grid: Nickel Grid, 300 mesh. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.