MilliporeSigma
HomeApplicationsProtein BiologyProtein Mass Spectrometry
Protein Mass Spectrometry
Protein mass spectrometry for protein identification, characterization, and quantitation
Protein mass spectrometry is widely used to analyze biological samples for biomarker discovery, proteomics research, and clinical applications. Compared to other techniques used for the large-scale characterization of proteins, mass spectrometry has become a primary tool for proteomics based on its amenability to complex analysis. 
Mass spectrometry is used to quantitatively identify and characterize proteins based on their structure, post-translational modifications, and interactions.  
  • Protein identification typically involves chemical or enzymatic digestion of proteins into peptides, which are then analyzed by mass spectrometry and identified using computational methods or sequencing.
  • Post-translational modifications can be identified through changes in amino acid residue mass. Sites of modification can be mapped using sequencing or computational methods.
  • For glycan analysis and profiling, enzymatic or chemical methods are used to release glycan moieties from glycoproteins, followed by derivatization of released glycans for mass spec analysis.
  • Protein interactions are determined by affinity co-purification of a specific target protein with any interacting proteins or studied more globally using size exclusion or ion exchange chromatography prior to analysis by mass spectrometry.