MilliporeSigma
HomeGel ElectrophoresisSetting Up and Running TruPAGE™ Precast Gels
Setting Up and Running TruPAGE™ Precast Gels
TruPAGE™ precast gels offer high rigidity for tear resistance and improved protein resolution so researchers can achieve quality separation the first time around. TruPAGE™ gel cassettes are compatible with popular gel electrophoresis equipment and have a hassle-free design, which includes extra tall wells to prevent lane to lane overflow, requires no tape and no comb removal, and does not require tools to open. Additionally, TruPAGE™ gels have an extended 2 year shelf life and are available at an affordable price compared to competitor's polyacrylamide gels.