MilliporeSigma
  • Gas chromatographic determination of alachlor in microencapsulated formulations: mini-collaborative study.
Gas chromatographic determination of alachlor in microencapsulated formulations: mini-collaborative study.
Journal - Association of Official Analytical Chemists (1987-11-01)
D F Tomkins
ABSTRACT
An isothermal gas chromatographic method for measuring alachlor in Micro-Tech (microencapsulated) formulations was tested by 5 collaborators. The samples were prepared in acetone, and alachlor was determined using a gas chromatographic column of 10% SP-2250 on 100-120 mesh Supelcoport. Di-n-pentylphthalate was used as the internal standard. Collaborators made single determinations on 5 samples distributed as blind duplicates. The mini-collaborative study generated 47 data points. The coefficient of variation (CVo-pooled) was 1.35%, and CVx-pooled was 0.73%. The method was simple to use and did not reveal any interferences in samples tested. The method has been adopted official first action as an AOAC-CIPAC method.