MilliporeSigma
  • 5-/12-Lipoxygenase-linked cascade contributes to the IL-33-induced synthesis of IL-13 in mast cells, thus promoting asthma development.
5-/12-Lipoxygenase-linked cascade contributes to the IL-33-induced synthesis of IL-13 in mast cells, thus promoting asthma development.
Allergy (2017-09-01)
MyungJa Ro, A-Jin Lee, Jae-Hong Kim
ABSTRACT
As asthma progresses, the levels of IL-33 in serum are markedly increased and contribute to asthmatic development and exacerbation. Mast cells, one of the principal effector cells in the pathogenesis of asthma, express high levels of the IL-33 receptor ST2 and have been shown to be activated by IL-33. Thus, IL-33 stimulates mast cells to produce Th2-type cytokines such as IL-13, thus contributing to asthmatic development. However, the signaling mechanism for IL-33-induced synthesis of Th2 cytokines, particularly IL-13, has not been fully elucidated in mast cells. The role of 5- or 12-LO in the IL-33-induced synthesis of IL-13 was investigated using knockdown or pharmacological inhibitors in bone marrow-derived mast cells (BMMCs) and animal model. Blockade of 5- or 12-LO significantly suppressed IL-33-induced synthesis of IL-13 in BMMCs. The subsequent action of 5- and 12-LO metabolites through their specific receptor, BLT2, was also critical for IL-33-induced synthesis of IL-13. We also demonstrated that the MyD88-p38 kinase cascade lies upstream of 5-/12-LO and that NF-κB lies downstream of 5-/12-LO to mediate the IL-33-induced synthesis of IL-13 in mast cells. Consistent with these findings, we observed that in an IL-33-administered asthmatic airway inflammation model, IL-13 levels were markedly increased in bronchoalveolar lavage fluid, but its levels were markedly suppressed by treatment with inhibitors of 5-LO, 12-LO or BLT2, further suggesting roles of 5-/12-LO in IL-33-induced IL-13 production. Our results suggest that "MyD88-5-/12-LO-BLT2-NF-κB" cascade significantly contributes to the IL-33-induced synthesis of IL-13 in mast cells, thus potentially contributing to asthmatic development and exacerbation.
Sigma-Aldrich
12(S)-Hydroxy-(5Z,8Z,10E,14Z)-eicosatetraenoic acid, ~100 μg/mL in ethanol, ≥95% (HPLC)