MilliporeSigma
  • Nascent DNA Proteomics Reveals a Chromatin Remodeler Required for Topoisomerase I Loading at Replication Forks.
Nascent DNA Proteomics Reveals a Chromatin Remodeler Required for Topoisomerase I Loading at Replication Forks.
Cell reports (2016-04-07)
Cyril Ribeyre, Ralph Zellweger, Maeva Chauvin, Nicole Bec, Christian Larroque, Massimo Lopes, Angelos Constantinou
ABSTRACT
During transcription and DNA replication, the DNA template is overwound ahead of RNA and DNA polymerases and relaxed by DNA topoisomerases. Inhibitors of topoisomerases are potent anti-cancer agents. Camptothecin traps topoisomerase I on DNA and exerts preferential cytotoxicity toward cancer cells by way of its interference with the progression of replication forks. Starting with an unbiased proteomic analysis, we find that the chromatin remodeling complex BAZ1B-SMARCA5 accumulates near replication forks in camptothecin-exposed cells. We report that BAZ1B associates with topoisomerase I and facilitates its access to replication forks. Single-molecule analyses of replication structures show that BAZ1B contributes to replication interference by camptothecin. A lack of BAZ1B confers increased cellular tolerance of camptothecin. These findings reveal BAZ1B as a key facilitator of topoisomerase I function during DNA replication that affects the response of cancer cells to topoisomerase I inhibitors.
Sigma-Aldrich
Normal Rabbit IgG, This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.
Sigma-Aldrich
Monoclonal Anti-Proliferating Cell Nuclear Antigen antibody produced in mouse, clone PC 10, ascites fluid