MilliporeSigma
  • X-band Electron Paramagnetic Resonance Investigation of Stable Organic Radicals Present under Cold Stratification in 'Fuji' Apple Seeds.
X-band Electron Paramagnetic Resonance Investigation of Stable Organic Radicals Present under Cold Stratification in 'Fuji' Apple Seeds.
Journal of oleo science (2017-12-05)
Kouichi Nakagawa, Kazuhiro Matsumoto, Nattakan Chaiserm, Aroonsri Priprem
ABSTRACT
We investigated stable organic radicals formed in response to cold stratification in 'Fuji' apple seeds using X-band (9 GHz) electron paramagnetic resonance (EPR) technique. This technique primarily detected two paramagnetic species in each seed. These two different radical species were assigned as a stable organic radical and Mn
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidinol, 98%