MilliporeSigma
  • Development of a thermo-stable and recyclable magnetic nanobiocatalyst for bioprocessing of fruit processing residues and D-allulose synthesis.
Development of a thermo-stable and recyclable magnetic nanobiocatalyst for bioprocessing of fruit processing residues and D-allulose synthesis.
Bioresource technology (2017-10-11)
Satya Narayan Patel, Vishal Singh, Manisha Sharma, Rajender S Sangwan, Nitin K Singhal, Sudhir P Singh
ABSTRACT
The aim of the study was to covalently immobilize Smt3-D-psicose 3-epimerase onto functionalized iron oxide magnetic nanoparticles. After immobilization, K
Sigma-Aldrich
D-Psicose, ≥95% (HPLC)
Sigma-Aldrich
3-Phosphonopropionic acid, technical grade, 94%