MilliporeSigma
  • Anti-aggregation effect of aroxyalkyl derivatives of 2-methoxyphenylpiperazine is due to their 5-HT
Anti-aggregation effect of aroxyalkyl derivatives of 2-methoxyphenylpiperazine is due to their 5-HT
European journal of pharmacology (2017-11-08)
Monika Kubacka, Grzegorz Kazek, Magdalena Kotańska, Barbara Filipek, Anna Maria Waszkielewicz, Szczepan Mogilski
ABSTRACT
Serotonin (5-HT) and adrenaline acting at platelet 5-HT
Sigma-Aldrich
Sarpogrelate hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Serotonin hydrochloride, powder