MilliporeSigma
  • Th17 Cells Induce Dopaminergic Neuronal Death via LFA-1/ICAM-1 Interaction in a Mouse Model of Parkinson's Disease.
Th17 Cells Induce Dopaminergic Neuronal Death via LFA-1/ICAM-1 Interaction in a Mouse Model of Parkinson's Disease.
Molecular neurobiology (2016-11-16)
Zhan Liu, Yan Huang, Bei-Bei Cao, Yi-Hua Qiu, Yu-Ping Peng
ABSTRACT
T helper (Th)17 cells, a subset of CD4
Sigma-Aldrich
Triton X-100, laboratory grade