MilliporeSigma
  • Pharmacokinetics, biodistribution and brain retention of a bispecific antibody-based PET radioligand for imaging of amyloid-β.
Pharmacokinetics, biodistribution and brain retention of a bispecific antibody-based PET radioligand for imaging of amyloid-β.
Scientific reports (2017-12-10)
Dag Sehlin, Xiaotian T Fang, Silvio R Meier, Malin Jansson, Stina Syvänen
ABSTRACT
Monoclonal antibodies (mAbs) have not been used as positron emission tomography (PET) ligands for in vivo imaging of the brain because of their limited passage across the blood-brain barrier (BBB). However, due to their high affinity and specificity, mAbs may be an attractive option for brain PET if their brain distribution can be facilitated. In the present study, a F(ab')
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)