MilliporeSigma
  • Novel Small Molecule Inhibitors of Protein Kinase D Suppress NF-kappaB Activation and Attenuate the Severity of Rat Cerulein Pancreatitis.
Novel Small Molecule Inhibitors of Protein Kinase D Suppress NF-kappaB Activation and Attenuate the Severity of Rat Cerulein Pancreatitis.
Frontiers in physiology (2017-12-23)
Jingzhen Yuan, Tanya Tan, Meng Geng, Grace Tan, Chintan Chheda, Stephen J Pandol
ABSTRACT
Nuclear factor-kappa B (NF-κB) activation is a key early signal regulating inflammatory and cell death responses in acute pancreatitis. Our previous
Sigma-Aldrich
GF 109203X, synthetic, ≥90% (HPLC)