MilliporeSigma
  • Kallikrein-related peptidase 4 induces cancer-associated fibroblast features in prostate-derived stromal cells.
Kallikrein-related peptidase 4 induces cancer-associated fibroblast features in prostate-derived stromal cells.
Molecular oncology (2017-05-17)
Thomas Kryza, Lakmali M Silva, Nathalie Bock, Ruth A Fuhrman-Luck, Carson R Stephens, Jin Gao, Hema Samaratunga, Mitchell G Lawrence, John D Hooper, Ying Dong, Gail P Risbridger, Judith A Clements
ABSTRACT
The reciprocal communication between cancer cells and their microenvironment is critical in cancer progression. Although involvement of cancer-associated fibroblasts (CAF) in cancer progression is long established, the molecular mechanisms leading to differentiation of CAFs from normal fibroblasts are poorly understood. Here, we report that kallikrein-related peptidase-4 (KLK4) promotes CAF differentiation. KLK4 is highly expressed in prostate epithelial cells of premalignant (prostatic intraepithelial neoplasia) and malignant lesions compared to normal prostate epithelia, especially at the peristromal interface. KLK4 induced CAF-like features in the prostate-derived WPMY1 normal stromal cell line, including increased expression of alpha-smooth muscle actin, ESR1 and SFRP1. KLK4 activated protease-activated receptor-1 in WPMY1 cells increasing expression of several factors (FGF1, TAGLN, LOX, IL8, VEGFA) involved in prostate cancer progression. In addition, KLK4 induced WPMY1 cell proliferation and secretome changes, which in turn stimulated HUVEC cell proliferation that could be blocked by a VEGFA antibody. Importantly, the genes dysregulated by KLK4 treatment of WPMY1 cells were also differentially expressed between patient-derived CAFs compared to matched nonmalignant fibroblasts and were further increased by KLK4 treatment. Taken together, we propose that epithelial-derived KLK4 promotes tumour progression by actively promoting CAF differentiation in the prostate stromal microenvironment.
Sigma-Aldrich
Human FGF1 / Fibroblast Growth Factor 1 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Ser-Leu-Ile-Gly-Lys-Val-amide, ≥95% (HPLC), solid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-TAGLN antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution