MilliporeSigma
  • Advanced glycation end products reduce the calcium transient in cardiomyocytes by increasing production of reactive oxygen species and nitric oxide.
Advanced glycation end products reduce the calcium transient in cardiomyocytes by increasing production of reactive oxygen species and nitric oxide.
FEBS open bio (2017-11-11)
Zeinab Hegab, Tamer M A Mohamed, Nicholas Stafford, Mamas Mamas, Elizabeth J Cartwright, Delvac Oceandy
ABSTRACT
Advanced glycation end products (AGE) are central to the development of cardiovascular complications associated with diabetes mellitus. AGE may alter cellular function through cross-linking of cellular proteins or by activating the AGE receptor (RAGE). However, the signalling molecules involved during AGE stimulation in cardiomyocytes remain unclear. Here, we investigated the effects of AGE treatment on intracellular calcium homeostasis of isolated cardiomyocytes and studied the activation of signalling molecules involved in this process. Treatment of cardiomyocytes with AGE for 24 h resulted in a dose-dependent reduction in calcium transient amplitude, reaching a maximum 50% reduction at a dose of 1 mg·mL