MilliporeSigma
RPE65 has an additional function as the lutein to
Proceedings of the National Academy of Sciences of the United States of America (2017-09-07)
Rajalekshmy Shyam, Aruna Gorusupudi, Kelly Nelson, Martin P Horvath, Paul S Bernstein
ABSTRACT
Carotenoids are plant-derived pigment molecules that vertebrates cannot synthesize de novo that protect the fovea of the primate retina from oxidative stress and light damage.