MilliporeSigma
  • Cardiovascular homeostasis dependence on MICU2, a regulatory subunit of the mitochondrial calcium uniporter.
Cardiovascular homeostasis dependence on MICU2, a regulatory subunit of the mitochondrial calcium uniporter.
Proceedings of the National Academy of Sciences of the United States of America (2017-10-27)
Alexander G Bick, Hiroko Wakimoto, Kimberli J Kamer, Yasemin Sancak, Olga Goldberger, Anna Axelsson, Daniel M DeLaughter, Joshua M Gorham, Vamsi K Mootha, J G Seidman, Christine E Seidman
ABSTRACT
Comparative analyses of transcriptional profiles from humans and mice with cardiovascular pathologies revealed consistently elevated expression of