MilliporeSigma
  • AFF1 and AFF4 differentially regulate the osteogenic differentiation of human MSCs.
AFF1 and AFF4 differentially regulate the osteogenic differentiation of human MSCs.
Bone research (2017-09-29)
Chen-Chen Zhou, Qiu-Chan Xiong, Xin-Xing Zhu, Wen Du, Peng Deng, Xiao-Bing Li, Yi-Zhou Jiang, Shu-Juan Zou, Cun-Yu Wang, Quan Yuan
ABSTRACT
AFF1 and AFF4 belong to the AFF (AF4/FMR2) family of proteins, which function as scaffolding proteins linking two different transcription elongation factors, positive elongation factor b (P-TEFb) and ELL1/2, in super elongation complexes (SECs). Both AFF1 and AFF4 regulate gene transcription through elongation and chromatin remodeling. However, their function in the osteogenic differentiation of mesenchymal stem cells (MSCs) is unknown. In this study, we show that small interfering RNA (siRNA)-mediated depletion of AFF1 in human MSCs leads to increased alkaline phosphatase (ALP) activity, enhanced mineralization and upregulated expression of osteogenic-related genes. On the contrary, depletion of AFF4 significantly inhibits the osteogenic potential of MSCs. In addition, we confirm that overexpression of AFF1 and AFF4 differentially affects osteogenic differentiation