MilliporeSigma
  • Identification of a novel YAP-14-3-3ζ negative feedback loop in gastric cancer.
Identification of a novel YAP-14-3-3ζ negative feedback loop in gastric cancer.
Oncotarget (2017-10-27)
Bin Zhang, Aihua Gong, Hui Shi, Qingli Bie, Zhaofeng Liang, Peipei Wu, Fei Mao, Hui Qian, Wenrong Xu
ABSTRACT
Growing evidence indicates that 14-3-3ζ and yes-associated protein (YAP) substantially promote tumorigenesis and tumor development. However, the regulatory mechanism underlying these two proteins remains unknown. Herein, we report a new regulatory role of 14-3-3ζ in the phosphorylation of YAP and the feedback inhibition of 14-3-3ζ by YAP. YAP and 14-3-3ζ expression exhibited a negative correlation in gastric cancer (GC) tissues. Moreover, patients with higher YAP and lower 14-3-3ζ expression had poor prognoses. Studies have revealed that 14-3-3ζ promotes cytoplasmic retention and suppresses the transcriptional activity of YAP by inducing its phosphorylation. Furthermore, we observed that the overexpression of YAP significantly reduced the expression of 14-3-3ζ by inducing its ubiquitination. YAP, 14-3-3ζ, and mouse double minute 2 homolog (MDM2) were colocalized, and the knockdown of MDM2 by siRNA attenuated the YAP-induced decrease of 14-3-3ζ. The binding of 14-3-3ζ and MDM2 was also restrained when the expression of YAP was interfered. Our results indicated the presence of a 14-3-3ζ-YAP negative regulatory feedback loop, which has a crucial role in cell proliferation and survival and is a potential target for the clinical treatment of GC.
Sigma-Aldrich
MISSION® esiRNA, targeting human MDM2