MilliporeSigma
Recombinant
Frontiers in microbiology (2017-10-07)
Ning Xu, Xiaolei Liu, Bin Tang, Libo Wang, Hai N Shi, Pascal Boireau, Mingyuan Liu, Xue Bai
ABSTRACT
During parasite infection, serine protease inhibitors secreted by parasites play important roles in suppressing host defenses. However, the mechanism of immune regulation is unclear. In this study, a serpin gene from
Sigma-Aldrich
N-Succinyl-Ala-Ala-Ala-p-nitroanilide, elastase substrate