MilliporeSigma
  • A glass fibre membrane platform for ultra-sensitive detection of cardiac troponin T.
A glass fibre membrane platform for ultra-sensitive detection of cardiac troponin T.
The Analyst (2017-09-28)
Keum-Soo Song, Satish Balasaheb Nimse, Mukesh Digambar Sonawane, Yahui Lin, Zhou Zhou, Taisun Kim
ABSTRACT
A glass fibre membrane platform that allows quantification of circulating cTnT with a LoD of 0.87 pg mL
Sigma-Aldrich
Hemoglobin human, lyophilized powder