MilliporeSigma
  • Disruption of Aneuploidy and Senescence Induced by Aurora Inhibition Promotes Intrinsic Apoptosis in Double Hit or Double Expressor Diffuse Large B-cell Lymphomas.
Disruption of Aneuploidy and Senescence Induced by Aurora Inhibition Promotes Intrinsic Apoptosis in Double Hit or Double Expressor Diffuse Large B-cell Lymphomas.
Molecular cancer therapeutics (2017-06-16)
Shariful Islam, Wenqing Qi, Carla Morales, Laurence Cooke, Catherine Spier, Eric Weterings, Daruka Mahadevan
ABSTRACT
Double hit (DH) or double expressor (DE) diffuse large B-cell lymphomas (DLBCL) are aggressive non-Hodgkin's lymphomas (NHL) with translocations and/or overexpressions of