MilliporeSigma
  • Clostridium difficile chimeric toxin receptor binding domain vaccine induced protection against different strains in active and passive challenge models.
Clostridium difficile chimeric toxin receptor binding domain vaccine induced protection against different strains in active and passive challenge models.
Vaccine (2017-07-04)
Jing-Hui Tian, Gregory Glenn, David Flyer, Bin Zhou, Ye Liu, Eddie Sullivan, Hua Wu, James F Cummings, Larry Elllingsworth, Gale Smith
ABSTRACT
Clostridium difficile is the number one cause of nosocomial antibiotic-associated diarrhea in developed countries. Historically, pathogenesis was attributed two homologous glucosylating toxins, toxin-A (TcdA) and toxin-B (TcdB). Over the past decade, however, highly virulent epidemic strains of C. difficile (B1/NAP1/027) have emerged and are linked to an increase in morbidity and mortality. Increased virulence is attributed to multiple factors including: increased production of A- and B-toxins; production of binary toxin (CDT); and the emergence of more toxic TcdB variants (TcdB
Sigma-Aldrich
1,1′-Carbonyl-di-(1,2,4-triazole), technical, ≥90% (T)