MilliporeSigma
  • Direct inhibition of STAT signaling by platinum drugs contributes to their anti-cancer activity.
Direct inhibition of STAT signaling by platinum drugs contributes to their anti-cancer activity.
Oncotarget (2017-09-15)
Stanleyson V Hato, Carl G Figdor, Susumu Takahashi, Anja E Pen, Altuna Halilovic, Kalijn F Bol, Angela Vasaturo, Yukie Inoue, Nienke de Haas, Dagmar Verweij, Carla M L Van Herpen, Johannes H Kaanders, Johan H J M van Krieken, Hanneke W M Van Laarhoven, Gerrit K J Hooijer, Cornelis J A Punt, Akira Asai, I Jolanda M de Vries, W Joost Lesterhuis
ABSTRACT
Platinum-based chemotherapeutics are amongst the most powerful anti-cancer drugs. Although their exact mechanism of action is not well understood, it is thought to be mediated through covalent DNA binding. We investigated the effect of platinum-based chemotherapeutics on signaling through signal transducer and activator of transcription (STAT) proteins, which are involved in many oncogenic signaling pathways. We performed