MilliporeSigma
  • The effects of fructose and phosphate infusions on dry matter intake of lactating cows.
The effects of fructose and phosphate infusions on dry matter intake of lactating cows.
Journal of dairy science (2017-01-31)
Roni Yair, Michael S Allen
ABSTRACT
The objective of this study was to examine the effects of fructose and phosphate (Pi) infusions on dry matter intake by dairy cows to further understand the mechanisms controlling feed intake related to hepatic energy status. We performed 3 experiments in which we infused fructose and Pi intravenously or abomasally to Holstein cows. The first experiment used 8 cows (4-8 d postpartum) in a duplicated 4 × 4 Latin square experiment with 1 square of multiparous and 1 square of primiparous cows. A 2 × 2 factorial arrangement of treatments was used including jugular infusions of solutions (1 L/h) containing fructose or glucose (0.6 mol/h) and Pi (NaH
Sigma-Aldrich
PGO Enzyme Preparation, 1 G capsules