MilliporeSigma
  • Changes in phagocytosis and potassium channel activity in microglia of 5xFAD mice indicate alterations in purinergic signaling in a mouse model of Alzheimer's disease.
Changes in phagocytosis and potassium channel activity in microglia of 5xFAD mice indicate alterations in purinergic signaling in a mouse model of Alzheimer's disease.
Neurobiology of aging (2017-07-12)
Stefan Wendt, Meron Maricos, Natascha Vana, Niklas Meyer, Dilansu Guneykaya, Marcus Semtner, Helmut Kettenmann
ABSTRACT
As the immunocompetent cells of the central nervous system, microglia accumulate at amyloid beta plaques in Alzheimer's disease (AD) and acquire a morphological phenotype of activated microglia. Recent functional studies, however, indicate that in mouse models of amyloidosis and AD, these cells are rather dysfunctional indicated by a reduced phagocytic activity. Here, we report that this reduction in phagocytic activity is associated with perturbed purinergic receptor signaling, since phagocytosis could be stimulated by P2Y
Sigma-Aldrich
Sulforhodamine 101, Dye content ~95 %