MilliporeSigma
  • Compound A influences gene regulation of the Dexamethasone-activated glucocorticoid receptor by alternative cofactor recruitment.
Compound A influences gene regulation of the Dexamethasone-activated glucocorticoid receptor by alternative cofactor recruitment.
Scientific reports (2017-08-16)
S J Desmet, N Bougarne, L Van Moortel, L De Cauwer, J Thommis, M Vuylsteke, D Ratman, R Houtman, J Tavernier, K De Bosscher
ABSTRACT
The glucocorticoid receptor (GR) is a transcription factor of which the underlying gene regulatory mechanisms are complex and incompletely understood. The non-steroidal anti-inflammatory Compound A (CpdA), a selective GR modulating compound in various cell models, has been shown to favour GR-mediated gene repression but not GR-mediated gene activation. Shifting balances towards only a particular subset of GR gene regulatory events may be of benefit in the treatment of inflammatory diseases. We present evidence to support that the combination of CpdA with Dexamethasone (DEX), a classic steroidal GR ligand, can shape GR function towards a unique gene regulatory profile in a cell type-dependent manner. The molecular basis hereof is a changed GR phosphorylation status concomitant with a change in the GR cofactor recruitment profile. We subsequently identified and confirmed the orphan nuclear receptor SHP as a coregulator that is specifically enriched at GR when CpdA and DEX are combined. Combining CpdA with DEX not only leads to stronger suppression of pro-inflammatory gene expression, but also enhanced anti-inflammatory GR target gene expression in epithelial cells, making ligand combination strategies in future a potentially attractive alternative manner of skewing and fine-tuning GR effects towards an improved therapeutic benefit.