MilliporeSigma
  • The Role of Calcium-Sensing Receptors in Endothelin-1-Dependent Effects on Adult Rat Ventricular Cardiomyocytes: Possible Contribution to Adaptive Myocardial Hypertrophy.
The Role of Calcium-Sensing Receptors in Endothelin-1-Dependent Effects on Adult Rat Ventricular Cardiomyocytes: Possible Contribution to Adaptive Myocardial Hypertrophy.
Journal of cellular physiology (2016-09-24)
Elena Dyukova, Rolf Schreckenberg, Christoph Arens, Guzel Sitdikova, Klaus-Dieter Schlüter
ABSTRACT
Nitric oxide (NO)-deficiency as it occurs during endothelial dysfunction activates the endothelin-1 (ET-1) system and increases the expression of receptor activity modifying protein (RAMP)-1 that acts as a chaperon for calcium-sensing receptors (CaR) that have recently been identified to improve cardiac function. Here, we hypothesized that ET-1 increases the cardiac expression of CaR and thereby induces an adaptive type of hypertrophy. Expressions of RAMP-1, endothelin receptors, and CaR were analyzed by RT-PCR in left ventricular tissues of L-NAME-treated rats. Effects of ET-1 on CaR expression and cell function (load free cell shortening) were analyzed in adult rat ventricular cardiomyocytes. siRNA directed against CaR and RAMP-1 was used to investigate a causal relationship. PD142893 and BQ788 were used to dissect the contribution of ET
Sigma-Aldrich
Anti-Calcium Sensing Receptor antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®