MilliporeSigma
  • XWL-1-48 exerts antitumor activity via targeting topoisomerase II and enhancing degradation of Mdm2 in human hepatocellular carcinoma.
XWL-1-48 exerts antitumor activity via targeting topoisomerase II and enhancing degradation of Mdm2 in human hepatocellular carcinoma.
Scientific reports (2017-09-01)
Yajie Wang, Hua Sun, Zhiyan Xiao, Dan Zhang, Xiuqi Bao, Ning Wei
ABSTRACT
A novel podophyllotoxin derivative, XWL-1-48, was synthesized as an oral topoisomerase II inhibitor. kDNA decatenation assay indicated that XWL-1-48 significantly inhibited topoisomerase II activity in a concentration-dependent manner. Moreover, the cytotoxicity of XWL-1-48 is more potent than its congener GL331 and the IC