MilliporeSigma
  • Non-canonical pathway induced by Wnt3a regulates β-catenin via Pyk2 in differentiating human neural progenitor cells.
Non-canonical pathway induced by Wnt3a regulates β-catenin via Pyk2 in differentiating human neural progenitor cells.
Biochemical and biophysical research communications (2017-07-12)
Venkata Ajay Narendra Talabattula, Peter Morgan, Moritz J Frech, Adelinde M Uhrmacher, Ottmar Herchenröder, Brigitte M Pützer, Arndt Rolfs, Jiankai Luo
ABSTRACT
Wnt/β-catenin and Wnt/Ca