MilliporeSigma
  • Signaling Lymphocytic Activation Molecule Family Member 7 Engagement Restores Defective Effector CD8+ T Cell Function in Systemic Lupus Erythematosus.
Signaling Lymphocytic Activation Molecule Family Member 7 Engagement Restores Defective Effector CD8+ T Cell Function in Systemic Lupus Erythematosus.
Arthritis & rheumatology (Hoboken, N.J.) (2017-01-12)
Denis Comte, Maria P Karampetsou, Nobuya Yoshida, Katalin Kis-Toth, Vasileios C Kyttaris, George C Tsokos
ABSTRACT
Effector CD8+ T cell function is impaired in systemic lupus erythematosus (SLE) and is associated with a compromised ability to fight infections. Signaling lymphocytic activation molecule family member 7 (SLAMF7) engagement has been shown to enhance natural killer cell degranulation. This study was undertaken to characterize the expression and function of SLAMF7 on CD8+ T cell subsets isolated from the peripheral blood of SLE patients and healthy subjects. CD8+ T cell subset distribution, SLAMF7 expression, and expression of cytolytic enzymes (perforin, granzyme A [GzmA], and GzmB) on cells isolated from SLE patients and healthy controls were analyzed by flow cytometry. CD107a expression and interferon-γ (IFNγ) production in response to viral antigenic stimulation in the presence or absence of an anti-SLAMF7 antibody were assessed by flow cytometry. Antiviral cytotoxic activity in response to SLAMF7 engagement was determined using a flow cytometry-based assay. The distribution of CD8+ T cell subsets was altered in the peripheral blood of SLE patients, with a decreased effector cell subpopulation. Memory CD8+ T cells from SLE patients displayed decreased amounts of SLAMF7, a surface receptor that characterizes effector CD8+ T cells. Ligation of SLAMF7 increased CD8+ T cell degranulation capacity and the percentage of IFNγ-producing cells in response to antigen challenge in SLE patients and healthy controls. Moreover, SLAMF7 engagement promoted cytotoxic lysis of target cells in response to stimulation with viral antigens. CD8+ T cell activation in response to viral antigens is defective in SLE patients. Activation of SLAMF7 through a specific monoclonal antibody restores CD8+ T cell antiviral effector function to normal levels and thus represents a potential therapeutic option in SLE.