MilliporeSigma
  • Regeneration of peroxiredoxins by p53-regulated sestrins, homologs of bacterial AhpD.
Regeneration of peroxiredoxins by p53-regulated sestrins, homologs of bacterial AhpD.
Science (New York, N.Y.) (2004-04-24)
Andrei V Budanov, Anna A Sablina, Elena Feinstein, Eugene V Koonin, Peter M Chumakov
ABSTRACT
Acting as a signal, hydrogen peroxide circumvents antioxidant defense by overoxidizing peroxiredoxins (Prxs), the enzymes that metabolize peroxides. We show that sestrins, a family of proteins whose expression is modulated by p53, are required for regeneration of Prxs containing Cys-SO(2)H, thus reestablishing the antioxidant firewall. Sestrins contain a predicted redox-active domain homologous to AhpD, the enzyme catalyzing the reduction of a bacterial Prx, AhpC. Purified Hi95 (sestrin 2) protein supports adenosine triphosphate-dependent reduction of overoxidized PrxI in vitro, indicating that unlike AhpD, which is a disulfide reductase, sestrins are cysteine sulfinyl reductases. As modulators of peroxide signaling and antioxidant defense, sestrins constitute potential therapeutic targets.